missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoroaniline, 99%

Catalog No. AAA1367336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1367336 500 g
AAA1367314 25 g
AAA1367322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1367336 Supplier Thermo Scientific Chemicals Supplier No. A1367336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 367-25-9 | C6H5F2N | 129.11 g/mol

2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 367-25-9
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007648
InChI Key CEPCPXLLFXPZGW-UHFFFAOYSA-N
Synonym 2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine
PubChem CID 9709
IUPAC Name 2,4-difluoroaniline
SMILES NC1=CC=C(F)C=C1F

Specifications

Melting Point -8°C to -7°C
Density 1.281
Boiling Point 168°C to 170°C
Flash Point 62°C (143°F)
Odor Characteristic
Refractive Index 1.507
Quantity 500 g
UN Number UN2941
Beilstein 2802556
Merck Index 14,3147
Solubility Information Not miscible or difficult to mix.
Formula Weight 129.11
Percent Purity 99%
Chemical Name or Material 2,4-Difluoroaniline
Show More Show Less
Hazard Category H227-H302-H311+H331
Hazard Statement GHS H Statement
H311-H331-H373-H227-H302-H315-H319
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
EINECSNumber 206-687-5
RTECSNumber BX3680000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only