missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Difluoroacetophenone, 99%

Catalog No. AC246770100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC246770100 10 g
AC246770500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC246770100 Supplier Thermo Scientific Chemicals Supplier No. 246770100

CAS: 364-83-0 | C8H6F2O | 156.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 364-83-0
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.12
MDL Number MFCD00151261
InChI Key QEWHNJPLPZOEKU-UHFFFAOYSA-N
Synonym 2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213
PubChem CID 67770
IUPAC Name 1-(2,4-difluorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 80.0°C to 81.0°C (25.0 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.4890
Linear Formula F2C6H3COCH3
Quantity 10 g
Specific Gravity 1.23
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble/miscible in polar solvents,insoluble in non-polar solvents
Formula Weight 156.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2', 4'-Difluoroacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-667-6

RUO – Research Use Only