accessibility menu, dialog, popup

UserName

H3COFF

CAS RN 364-83-0

IUPAC Name: 1-(2,4-difluorophenyl)ethan-1-one
Synonyms: 1-(2,4-difluorophenyl)ethanone
Molecular Weight (g/mol): 156.13
Molecular Formula: C8H6F2O
InChi Key: QEWHNJPLPZOEKU-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(F)C=C1F
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)