missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-5-nitrophenol, 98%

Catalog No. AAH3208106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3208106 5 g
AAH3208114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3208106 Supplier Thermo Scientific Chemicals Supplier No. H3208106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 113512-57-5 | C6H3F2NO3 | 175.091 g/mol

2,4-Difluoro-5-nitrophenol may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoro-5-nitrophenol may be used in chemical synthesis.

Solubility
Sparingly soluble in water but readily soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 113512-57-5
Molecular Formula C6H3F2NO3
Molecular Weight (g/mol) 175.091
MDL Number MFCD08443942
InChI Key SMRYCTJAGPDVEH-UHFFFAOYSA-N
Synonym 2,4-difluoro-5-nitrobenzenol, phenol, 2,4-difluoro-5-nitro, difluoronitrobenzenol, acmc-209wyr, 2,4-difluoro-5-hydroxynitrobenzene
PubChem CID 10921024
IUPAC Name 2,4-difluoro-5-nitrophenol
SMILES C1=C(C(=CC(=C1O)F)F)[N+](=O)[O-]

Specifications

Melting Point 112°C to 120°C
Quantity 5 g
Solubility Information Sparingly soluble in water but readily soluble in organic solvents.
Formula Weight 175.09
Percent Purity 98%
Chemical Name or Material 2,4-Difluoro-5-nitrophenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only