missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-5-nitrobenzoic acid, 97%

Catalog No. AAH3261703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3261703 1 g
AAH3261706 5 g
AAH3261714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3261703 Supplier Thermo Scientific Chemicals Supplier No. H3261703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 153775-33-8 | C7H2F2NO4 | 202.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 153775-33-8
Molecular Formula C7H2F2NO4
Molecular Weight (g/mol) 202.09
MDL Number MFCD04115713
InChI Key MDFSGDMPHMKKGB-UHFFFAOYSA-M
PubChem CID 2782822
IUPAC Name 2,4-difluoro-5-nitrobenzoate
SMILES [O-]C(=O)C1=CC(=C(F)C=C1F)[N+]([O-])=O

Specifications

Melting Point 125°C to 133°C
Quantity 1 g
Synonym 2,4-difluoro-5-nitrobenzoic acid, benzoic acid, 2,4-difluoro-5-nitro, benzoic acid,2,4-difluoro-5-nitro, 2,4-difluoro-5-nitro-benzoic acid, acmc-209d9v, ksc527s6t, 2,4-difluoro-5-nitrobenzoicacid, buttpark 156\01-93
Formula Weight 203.1
Percent Purity 97%
Chemical Name or Material 2,4-Difluoro-5-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only