missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. D53091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D53091G Supplier TCI America Supplier No. D53091G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 178974-97-5
Melting Point 195°C
Color White-Yellow
Molecular Formula C8H6F2O3
Quantity 1 g
MDL Number MFCD02258905
Synonym 2,4-difluoro-3-methoxy-benzoic acid, 2,4-difluoro-3-methoxylbenzoic acid, 2,4-difluoro-3-methoxy benzoic acid, pubchem4941, acmc-1cabj, 3-methoxy-2,4-difluorobenzoic acid, benzoic acid,2,4-difluoro-3-methoxy
InChI Key KWLIGHXPUYUTBH-UHFFFAOYSA-N
SMILES COC1=C(F)C=CC(C(O)=O)=C1F
IUPAC Name 2,4-difluoro-3-methoxybenzoic acid
Molecular Weight (g/mol) 188.13
PubChem CID 3780555
Formula Weight 188.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Difluoro-3-methoxybenzoic Acid
Show More Show Less