missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-methoxybenzoic acid, 98+%

Catalog No. AAB2559706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2559706 5 g
AAB2559714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2559706 Supplier Thermo Scientific Chemicals Supplier No. B2559706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 178974-97-5 | C8H6F2O3 | 188.13 g/mol

2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoro-3-methoxybenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 178974-97-5
Molecular Formula C8H6F2O3
Molecular Weight (g/mol) 188.13
MDL Number MFCD02258905
InChI Key KWLIGHXPUYUTBH-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-methoxy-benzoic acid, 2,4-difluoro-3-methoxylbenzoic acid, 2,4-difluoro-3-methoxy benzoic acid, pubchem4941, acmc-1cabj, 3-methoxy-2,4-difluorobenzoic acid, benzoic acid,2,4-difluoro-3-methoxy
PubChem CID 3780555
IUPAC Name 2,4-difluoro-3-methoxybenzoic acid
SMILES COC1=C(F)C=CC(C(O)=O)=C1F

Specifications

Melting Point 193°C to 195°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 188.13
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluoro-3-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only