missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-1-iodobenzene, 99%

Catalog No. p-7081004
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1079809 10 g
AAA1079818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1079809 Supplier Thermo Scientific Chemicals Supplier No. A1079809

CAS: 2265-93-2 | C6H3F2I | 239.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2265-93-2
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.99
MDL Number MFCD00040151
InChI Key YKLDMAPEGQYZRT-UHFFFAOYSA-N
Synonym 2,4-difluoroiodobenzene, 1,3-difluoro-4-iodobenzene, 2,4-difluoro-1-iodobenze, benzene, 2,4-difluoro-1-iodo, 2,4-difluoro iodobenzene, 2,4-difluoro-1-iodo-benzene, pubchem3463, 2,4-difluoro-iodobenzene, acmc-1cq0v, intermediates-zcf02607
PubChem CID 137519
IUPAC Name 2,4-difluoro-1-iodobenzene
SMILES FC1=CC=C(I)C(F)=C1

Specifications

Density 2.006
Boiling Point 175°C to 176°C
Flash Point 67°C (152°F)
Refractive Index 1.558
Quantity 10 g
Beilstein 2081078
Sensitivity Light sensitive
Formula Weight 239.99
Percent Purity 99%
Chemical Name or Material 2,4-Difluoro-1-iodobenzene
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only