missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic Acid 98.0+%, TCI America™
$96.26 - $284.92
Chemical Identifiers
| CAS | 5809-23-4 |
|---|---|
| Molecular Formula | C18H19NO4 |
| Molecular Weight (g/mol) | 313.353 |
| MDL Number | MFCD00134669 |
| InChI Key | FQNKTJPBXAZUGC-UHFFFAOYSA-N |
| PubChem CID | 79878 |
| IUPAC Name | 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoic acid |
| SMILES | CCN(CC)C1=CC(=C(C=C1)C(=O)C2=CC=CC=C2C(=O)O)O |
Chemical Identifiers
| 5809-23-4 | |
| 313.353 | |
| FQNKTJPBXAZUGC-UHFFFAOYSA-N | |
| 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoic acid |
| C18H19NO4 | |
| MFCD00134669 | |
| 79878 | |
| CCN(CC)C1=CC(=C(C=C1)C(=O)C2=CC=CC=C2C(=O)O)O |
Specifications
| 5809-23-4 | |
| C18H19NO4 | |
| 25 g | |
| FQNKTJPBXAZUGC-UHFFFAOYSA-N | |
| 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoic acid | |
| 79878 | |
| ≥98.0% (GC,T) | |
| 2-(4-Diethylamino-2-hydroxybenzoyl)benzoic Acid |
| 180°C | |
| MFCD00134669 | |
| 2-Hydroxy-4-diethylamino-2′C-hydroxycarbonylbenzophenone | |
| CCN(CC)C1=CC(=C(C=C1)C(=O)C2=CC=CC=C2C(=O)O)O | |
| 313.353 | |
| 313.35 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0148
TSCA : Yes