missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Diethylamino-2-hydroxybenzoyl)benzoic Acid 98.0+%, TCI America™

Catalog No. D2140100G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2140100G 100 g
D214025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2140100G Supplier TCI America Supplier No. D2140100G

Chemical Identifiers

CAS 5809-23-4
Molecular Formula C18H19NO4
Molecular Weight (g/mol) 313.353
MDL Number MFCD00134669
InChI Key FQNKTJPBXAZUGC-UHFFFAOYSA-N
PubChem CID 79878
IUPAC Name 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoic acid
SMILES CCN(CC)C1=CC(=C(C=C1)C(=O)C2=CC=CC=C2C(=O)O)O

Specifications

Melting Point 180°C
Quantity 100 g
Synonym 2-Hydroxy-4-diethylamino-2′-hydroxycarbonylbenzophenone
Formula Weight 313.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Diethylamino-2-hydroxybenzoyl)benzoic Acid
EINECSNumber (4)-0148
TSCA Yes