missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diethyl-1,5-pentanediol (DL- and meso- mixture) 93.0+%, TCI America™

Catalog No. p-7127405
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D261925ML 25 mL
D2619500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D261925ML Supplier TCI America Supplier No. D261925ML

Chemical Identifiers

CAS 57987-55-0
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.257
MDL Number MFCD01861285
InChI Key OJRJDENLRJHEJO-UHFFFAOYSA-N
PubChem CID 11344163
IUPAC Name 2,4-diethylpentane-1,5-diol
SMILES CCC(CC(CC)CO)CO

Specifications

Color Colorless
Boiling Point 150°C
Quantity 25 mL
Formula Weight 160.26
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Diethyl-1,5-pentanediol (DL- and meso- mixture)
EINECSNumber (2)-0240
TSCA Yes