accessibility menu, dialog, popup

UserName

H3CHOCH3OH

CAS RN 57987-55-0

IUPAC Name: 2,4-diethylpentane-1,5-diol
Synonyms: 2,4-diethylpentane-1,5-diol
Molecular Weight (g/mol): 160.26
Molecular Formula: C9H20O2
InChi Key: OJRJDENLRJHEJO-UHFFFAOYNA-N
SMILES: CCC(CO)CC(CC)CO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results