missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorotoluene 98.0+%, TCI America™

Catalog No. p-7126198
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
D041925ML 25 mL
D0419500ML 500 mL
2 options

Catalog No. D041925ML

Supplier: TCI America D041925ML

Only null left

Chemical Identifiers

CAS 95-73-8
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000583
InChI Key FUNUTBJJKQIVSY-UHFFFAOYSA-N
Synonym 2,4-dichlorotoluene, benzene, 2,4-dichloro-1-methyl, toluene, 2,4-dichloro, 2,4-dichloro-1-methyl-benzene, unii-3s32n6lg3x, 2,4-dichloromethylbenzene, 2,4-dct, 2,4-diclorotoluene, 4,6-dichlorotoluene, 2,4-dichloro-toluen
PubChem CID 7254
ChEBI CHEBI:81651
IUPAC Name 2,4-dichloro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Cl)Cl

Specifications

Color Colorless
Boiling Point 198°C
Quantity 25 mL
UN Number 3082
Formula Weight 161.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dichlorotoluene
EINECSNumber (3)-0078
RTECSNumber XT0730000
TSCA Yes