missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorotoluene 98.0+%, TCI America™

Catalog No. D041925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D041925ML 25 mL
D0419500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D041925ML Supplier TCI America Supplier No. D041925ML

Chemical Identifiers

CAS 95-73-8
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000583
InChI Key FUNUTBJJKQIVSY-UHFFFAOYSA-N
Synonym 2,4-dichlorotoluene, benzene, 2,4-dichloro-1-methyl, toluene, 2,4-dichloro, 2,4-dichloro-1-methyl-benzene, unii-3s32n6lg3x, 2,4-dichloromethylbenzene, 2,4-dct, 2,4-diclorotoluene, 4,6-dichlorotoluene, 2,4-dichloro-toluen
PubChem CID 7254
ChEBI CHEBI:81651
IUPAC Name 2,4-dichloro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Cl)Cl

Specifications

Color Colorless
Boiling Point 198°C
Quantity 25 mL
UN Number 3082
Formula Weight 161.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dichlorotoluene
EINECSNumber (3)-0078
RTECSNumber XT0730000
TSCA Yes