missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorotoluene, 99%

CAS: 95-73-8 | C7H6Cl2 | 161.03 g/mol

Supplier:  Thermo Scientific Chemicals 163280010

 View more versions of this product

Catalog No. AC163280010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 4-Dichlorotoluene
95-73-8
100.0
Colorless
200.0°C
Authentic
Glass bottle
1.5450 to 1.5470
MFCD00000583
05, 295
2,4-dichlorotoluene, benzene, 2,4-dichloro-1-methyl, toluene, 2,4-dichloro, 2,4-dichloro-1-methyl-benzene, unii-3s32n6lg3x, 2,4-dichloromethylbenzene, 2,4-dct, 2,4-diclorotoluene, 4,6-dichlorotoluene, 2,4-dichloro-toluen
FUNUTBJJKQIVSY-UHFFFAOYSA-N
2,4-dichloro-1-methylbenzene
7254
161.03
Liquid
99%
98.5
-14.0°C
1.2400g/mL
79°C
98.5% min. (GC)
C7H6Cl2
CH3C6H3Cl2
1 L
1.24
Solubility in water: immiscible. Other solubilities: soluble in alcohol,ether and acetone
CC1=C(C=C(C=C1)Cl)Cl
161.03
CHEBI:81651
99%

Chemical Identifiers

95-73-8
161.03
FUNUTBJJKQIVSY-UHFFFAOYSA-N
7254
2,4-dichloro-1-methylbenzene
C7H6Cl2
MFCD00000583
2,4-dichlorotoluene, benzene, 2,4-dichloro-1-methyl, toluene, 2,4-dichloro, 2,4-dichloro-1-methyl-benzene, unii-3s32n6lg3x, 2,4-dichloromethylbenzene, 2,4-dct, 2,4-diclorotoluene, 4,6-dichlorotoluene, 2,4-dichloro-toluen
CHEBI:81651
CC1=C(C=C(C=C1)Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 202-445-8

RTECSNumber : XT0730000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only