missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloropyrimidine 98.0+%, TCI America™
$67.38 - $196.36
Chemical Identifiers
| CAS | 3934-20-1 |
|---|---|
| Molecular Formula | C4H2Cl2N2 |
| Molecular Weight (g/mol) | 148.974 |
| MDL Number | MFCD00006061 |
| InChI Key | BTTNYQZNBZNDOR-UHFFFAOYSA-N |
| Synonym | pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine |
| PubChem CID | 77531 |
| IUPAC Name | 2,4-dichloropyrimidine |
| SMILES | C1=CN=C(N=C1Cl)Cl |
Chemical Identifiers
| 3934-20-1 | |
| 148.974 | |
| BTTNYQZNBZNDOR-UHFFFAOYSA-N | |
| 77531 | |
| C1=CN=C(N=C1Cl)Cl |
| C4H2Cl2N2 | |
| MFCD00006061 | |
| pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine | |
| 2,4-dichloropyrimidine |
Specifications
| 3934-20-1 | |
| White-Yellow | |
| C4H2Cl2N2 | |
| 5 g | |
| BTTNYQZNBZNDOR-UHFFFAOYSA-N | |
| 2,4-dichloropyrimidine | |
| 77531 | |
| ≥98.0% (GC) | |
| 2,4-Dichloropyrimidine |
| 60°C | |
| 101°C | |
| MFCD00006061 | |
| pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine | |
| C1=CN=C(N=C1Cl)Cl | |
| 148.974 | |
| 148.97 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes