missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloropyrimidine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 162650250

 View more versions of this product

Catalog No. AC162650250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 4-Dichloropyrimidine
96.0
101°C (23.0mmHg)
96% min. (GC)
C4H2Cl2N2
25g
pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine
C1=CN=C(N=C1Cl)Cl
148.98
148.98
3934-20-1
100.0
Authentic
Glass bottle
MFCD00006061
23, 90
BTTNYQZNBZNDOR-UHFFFAOYSA-N
2,4-dichloropyrimidine
77531
97%

Chemical Identifiers

3934-20-1
148.98
BTTNYQZNBZNDOR-UHFFFAOYSA-N
77531
C1=CN=C(N=C1Cl)Cl
C4H2Cl2N2
MFCD00006061
pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine
2,4-dichloropyrimidine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 223-508-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only