missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloropyrimidine, 97%

Catalog No. AC162650050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162650050 5 g Glass bottle
AC162650250 25 g Glass bottle
AC162651000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC162650050 Supplier Thermo Scientific Chemicals Supplier No. 162650050

CAS: 3934-20-1 | C4H2Cl2N2 | 148.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3934-20-1
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.98
MDL Number MFCD00006061
InChI Key BTTNYQZNBZNDOR-UHFFFAOYSA-N
PubChem CID 77531
IUPAC Name 2,4-dichloropyrimidine
SMILES C1=CN=C(N=C1Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 57°C to 62°C
Color Beige to White or Gray
Boiling Point 101°C (23.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 23, 90
Synonym pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine
Formula Weight 148.98
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2, 4-Dichloropyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-508-6

RUO – Research Use Only