missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4'-Dichloropropiophenone 97.0+%, TCI America™
Supplier: TCI America D39375G
Specifications
| 2′,4′-Dichloropropiophenone | |
| Yellow | |
| C9H8Cl2O | |
| 5 g | |
| FBMTWRZQBRHOPF-UHFFFAOYSA-N | |
| 1-(2,4-dichlorophenyl)propan-1-one | |
| 123467 | |
| ≥97.0% (GC) |
| 37885-41-9 | |
| 123°C | |
| MFCD00027396 | |
| 2',4'-dichloropropiophenone, 1-2,4-dichlorophenyl propan-1-one, 2,4-dichloropropiophenone, 1-2,4-dichlorophenyl-1-propanone, 1-propanone, 1-2,4-dichlorophenyl, 2 ,4-dichloropropiophenone, pubchem24408, acmc-209ivy, timtec-bb sbb016918, 1-propanone,1-2,4-dichlorophenyl | |
| CCC(=O)C1=C(C=C(C=C1)Cl)Cl | |
| 203.062 | |
| 203.06 | |
| Liquid |
Chemical Identifiers
| 37885-41-9 | |
| 203.062 | |
| FBMTWRZQBRHOPF-UHFFFAOYSA-N | |
| 123467 | |
| CCC(=O)C1=C(C=C(C=C1)Cl)Cl |
| C9H8Cl2O | |
| MFCD00027396 | |
| 2',4'-dichloropropiophenone, 1-2,4-dichlorophenyl propan-1-one, 2,4-dichloropropiophenone, 1-2,4-dichlorophenyl-1-propanone, 1-propanone, 1-2,4-dichlorophenyl, 2 ,4-dichloropropiophenone, pubchem24408, acmc-209ivy, timtec-bb sbb016918, 1-propanone,1-2,4-dichlorophenyl | |
| 1-(2,4-dichlorophenyl)propan-1-one |
Safety and Handling
TSCA : No