accessibility menu, dialog, popup

UserName

CAS RN 37885-41-9

IUPAC Name: 1-(2,4-dichlorophenyl)propan-1-one
Synonyms: 1-(2,4-dichlorophenyl)propan-1-one
Molecular Weight (g/mol): 203.06
Molecular Formula: C9H8Cl2O
InChi Key: FBMTWRZQBRHOPF-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results