missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. D49125G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D49125G Supplier TCI America Supplier No. D49125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 6590-96-1
Melting Point 42°C
Color White-Yellow
Molecular Formula C7H3Cl2NS
Quantity 5 g
MDL Number MFCD00037827
UN Number 1759
Synonym 2,4-dichlorophenyl isothiocyanate, 2,4-dichlorophenylisothiocyanate, benzene,2,4-dichloro-1-isothiocyanato, benzene, 2,4-dichloro-1-isothiocyanato, 2,4-dichlorobenzenisothiocyanate, acmc-1b3n1, tos-bb-1123, 2,4-dichloro-1-isothiocyanato-benzene, #
InChI Key WVBNZZHGECFCSH-UHFFFAOYSA-N
SMILES ClC1=CC=C(N=C=S)C(Cl)=C1
IUPAC Name 2,4-dichloro-1-isothiocyanatobenzene
Molecular Weight (g/mol) 204.07
PubChem CID 81066
Formula Weight 204.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dichlorophenyl Isothiocyanate
Show More Show Less