missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenyl isothiocyanate, 97%

CAS: 6590-96-1 | C7H3Cl2NS | 204.07 g/mol

$164.70 - $164.70

Chemical Identifiers

CAS 6590-96-1
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00037827
InChI Key WVBNZZHGECFCSH-UHFFFAOYSA-N
Synonym 2,4-dichlorophenyl isothiocyanate, 2,4-dichlorophenylisothiocyanate, benzene,2,4-dichloro-1-isothiocyanato, benzene, 2,4-dichloro-1-isothiocyanato, 2,4-dichlorobenzenisothiocyanate, acmc-1b3n1, tos-bb-1123, 2,4-dichloro-1-isothiocyanato-benzene, #
PubChem CID 81066
IUPAC Name 2,4-dichloro-1-isothiocyanatobenzene
SMILES ClC1=CC=C(N=C=S)C(Cl)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2017814
View Documents
Thermo Scientific Chemicals
B2017814
25 g
Each for $164.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6590-96-1
204.07
WVBNZZHGECFCSH-UHFFFAOYSA-N
81066
ClC1=CC=C(N=C=S)C(Cl)=C1
C7H3Cl2NS
MFCD00037827
2,4-dichlorophenyl isothiocyanate, 2,4-dichlorophenylisothiocyanate, benzene,2,4-dichloro-1-isothiocyanato, benzene, 2,4-dichloro-1-isothiocyanato, 2,4-dichlorobenzenisothiocyanate, acmc-1b3n1, tos-bb-1123, 2,4-dichloro-1-isothiocyanato-benzene, #
2,4-dichloro-1-isothiocyanatobenzene

Specifications

6590-96-1
1.41
>110°C (230°F)
C7H3Cl2NS
25 g
777072
2,4-dichlorophenyl isothiocyanate, 2,4-dichlorophenylisothiocyanate, benzene,2,4-dichloro-1-isothiocyanato, benzene, 2,4-dichloro-1-isothiocyanato, 2,4-dichlorobenzenisothiocyanate, acmc-1b3n1, tos-bb-1123, 2,4-dichloro-1-isothiocyanato-benzene, #
ClC1=CC=C(N=C=S)C(Cl)=C1
204.07
204.08
2,4-Dichlorophenyl isothiocyanate
36°C to 39°C
130°C to 133°C (2 mmHg)
Irritating
MFCD00037827
UN2923
Moisture sensitive
WVBNZZHGECFCSH-UHFFFAOYSA-N
2,4-dichloro-1-isothiocyanatobenzene
81066
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301+H311+H331-H314-H317-H334-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 229-527-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only