missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenyl isothiocyanate, 97%

Catalog No. AAB2017814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2017814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2017814 Supplier Thermo Scientific Chemicals Supplier No. B2017814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6590-96-1 | C7H3Cl2NS | 204.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6590-96-1
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00037827
InChI Key WVBNZZHGECFCSH-UHFFFAOYSA-N
Synonym 2,4-dichlorophenyl isothiocyanate, 2,4-dichlorophenylisothiocyanate, benzene,2,4-dichloro-1-isothiocyanato, benzene, 2,4-dichloro-1-isothiocyanato, 2,4-dichlorobenzenisothiocyanate, acmc-1b3n1, tos-bb-1123, 2,4-dichloro-1-isothiocyanato-benzene, #
PubChem CID 81066
IUPAC Name 2,4-dichloro-1-isothiocyanatobenzene
SMILES ClC1=CC=C(N=C=S)C(Cl)=C1

Specifications

Melting Point 36°C to 39°C
Density 1.41
Boiling Point 130°C to 133°C (2 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Quantity 25 g
UN Number UN2923
Beilstein 777072
Sensitivity Moisture sensitive
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2,4-Dichlorophenyl isothiocyanate
Show More Show Less
Hazard Category H301+H311+H331-H314-H317-H334-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 229-527-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only