missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenyl isocyanate, 96%

CAS: 2612-57-9 | C7H3Cl2NO | 188.007 g/mol

Supplier:  Thermo Scientific Chemicals L1108906

 View more versions of this product

Catalog No. AAL1108906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dichlorophenyl isocyanate
57°C to 61°C
Irritating
MFCD00002001
UN2250
Moisture sensitive
OLBJNSPBWLCTOT-UHFFFAOYSA-N
2,4-dichloro-1-isocyanatobenzene
75789
96%
2612-57-9
80°C (1 mmHg)
C7H3Cl2NO
5 g
2090780
2,4-dichlorophenyl isocyanate, 2,4-dichlorophenylisocyanate, benzene, 2,4-dichloro-1-isocyanato, 2,4-dichlorophenyl isocyantae, 2,4-dichlorobenzenisocyanate, acmc-1cg9n, 2,4-dichlorphenyl isocyanate, 2,4-dichloro phenyl isocyanate, #
C1=CC(=C(C=C1Cl)Cl)N=C=O
188.007
188.01

Chemical Identifiers

2612-57-9
188.007
OLBJNSPBWLCTOT-UHFFFAOYSA-N
75789
C1=CC(=C(C=C1Cl)Cl)N=C=O
C7H3Cl2NO
MFCD00002001
2,4-dichlorophenyl isocyanate, 2,4-dichlorophenylisocyanate, benzene, 2,4-dichloro-1-isocyanato, 2,4-dichlorophenyl isocyantae, 2,4-dichlorobenzenisocyanate, acmc-1cg9n, 2,4-dichlorphenyl isocyanate, 2,4-dichloro phenyl isocyanate, #
2,4-dichloro-1-isocyanatobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c

H301+H311+H331-H315-H317-H319-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROPHENYL ISOCYANATES

EINECSNumber : 220-040-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only