missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenyl isocyanate, 96%

Catalog No. p-7045540
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1108903 1 g
AAL1108906 5 g
AAL1108914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1108903 Supplier Thermo Scientific Chemicals Supplier No. L1108903

CAS: 2612-57-9 | C7H3Cl2NO | 188.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2612-57-9
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00002001
InChI Key OLBJNSPBWLCTOT-UHFFFAOYSA-N
Synonym 2,4-dichlorophenyl isocyanate, 2,4-dichlorophenylisocyanate, benzene, 2,4-dichloro-1-isocyanato, 2,4-dichlorophenyl isocyantae, 2,4-dichlorobenzenisocyanate, acmc-1cg9n, 2,4-dichlorphenyl isocyanate, 2,4-dichloro phenyl isocyanate, #
PubChem CID 75789
IUPAC Name 2,4-dichloro-1-isocyanatobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)N=C=O

Specifications

Melting Point 57°C to 61°C
Boiling Point 80°C (1 mmHg)
Odor Irritating
Quantity 1 g
UN Number UN2250
Beilstein 2090780
Sensitivity Moisture sensitive
Formula Weight 188.01
Percent Purity 96%
Chemical Name or Material 2,4-Dichlorophenyl isocyanate
Hazard Category H301+H311+H331-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROPHENYL ISOCYANATES
EINECSNumber 220-040-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only