missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenoxyacetic Acid 97.0+%, TCI America™

Catalog No. p-7126184
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D039625G 25 g
D0396500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D039625G Supplier TCI America Supplier No. D039625G
Only null left

Chemical Identifiers

CAS 94-75-7
Molecular Formula C8H6Cl2O3
Molecular Weight (g/mol) 221.033
MDL Number MFCD00004300
InChI Key OVSKIKFHRZPJSS-UHFFFAOYSA-N
Synonym 2,4-dichlorophenoxyacetic acid, 2-2,4-dichlorophenoxy acetic acid, 2,4-dichlorophenoxy acetic acid, hedonal, rhodia, agrotect, fernesta, fernimine, netagrone, tributon
PubChem CID 1486
ChEBI CHEBI:28854
IUPAC Name 2-(2,4-dichlorophenoxy)acetic acid
SMILES C1=CC(=C(C=C1Cl)Cl)OCC(=O)O

Specifications

Melting Point 139°C
Boiling Point 160°C
Quantity 25 g
UN Number 3077
Formula Weight 221.03
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorophenoxyacetic Acid
EINECSNumber (3)-0927
RTECSNumber AG6825000
TSCA Yes