missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenoxyacetic acid, 98%

CAS: 94-75-7 | C8H6Cl2O3 | 221.033 g/mol

Supplier:  Thermo Scientific Chemicals A1246730

 View more versions of this product

Catalog No. AAA1246730


Only null left
Explore available promotions

Description

Description

2,4-Dichlorophenoxyacetic acid is used as a supplement in plant cell culture media. It is a synthetic auxin, which finds application as a plant growth regulator. Further, it is used to increase latex output of old rubber trees.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorophenoxyacetic acid is used as a supplement in plant cell culture media. It is a synthetic auxin, which finds application as a plant growth regulator. Further, it is used to increase latex output of old rubber trees.

Solubility
Slightly soluble in water.

Notes
Moisture and light-sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

2,4-Dichlorophenoxyacetic acid
136°C to 140°C
160°C (0.4 mmHg)
C8H6Cl2O3
250 g
14,2796
Slightly soluble in water.
C1=CC(=C(C=C1Cl)Cl)OCC(=O)O
221.033
CHEBI:28854
98%
94-75-7
1.42
Odorless
MFCD00004300
1214242
2,4-dichlorophenoxyacetic acid, 2-2,4-dichlorophenoxy acetic acid, 2,4-dichlorophenoxy acetic acid, hedonal, rhodia, agrotect, fernesta, fernimine, netagrone, tributon
OVSKIKFHRZPJSS-UHFFFAOYSA-N
2-(2,4-dichlorophenoxy)acetic acid
1486
221.04

Chemical Identifiers

94-75-7
221.033
OVSKIKFHRZPJSS-UHFFFAOYSA-N
1486
2-(2,4-dichlorophenoxy)acetic acid
C8H6Cl2O3
MFCD00004300
2,4-dichlorophenoxyacetic acid, 2-2,4-dichlorophenoxy acetic acid, 2,4-dichlorophenoxy acetic acid, hedonal, rhodia, agrotect, fernesta, fernimine, netagrone, tributon
CHEBI:28854
C1=CC(=C(C=C1Cl)Cl)OCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H312-H317-H335
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c

H302-H317-H318-H335-H351

EINECSNumber : 202-361-1

RTECSNumber : AG6825000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only