missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorofluorobenzene 96.0+%, TCI America™

Catalog No. p-7127090
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D205925G 25 g
D2059500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D205925G Supplier TCI America Supplier No. D205925G
Only null left

Chemical Identifiers

CAS 1435-48-9
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.988
MDL Number MFCD00000580
InChI Key BDJZCCWUSOZUQG-UHFFFAOYSA-N
Synonym 1,3-dichloro-4-fluorobenzene, 2,4-dichlorofluorobenzene, benzene, 2,4-dichloro-1-fluoro, dcfb, benzene,2,4-dichloro-1-fluoro, 2,4-dichloro-1-fluoro-benzene, pubchem2312, acmc-209cqu, 2,4-dichlorofluobenzene, 1,3-dichlorofluorobenzene
PubChem CID 123112
IUPAC Name 2,4-dichloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)F

Specifications

Color Yellow
Boiling Point 173°C
Quantity 25 g
UN Number 1993
Formula Weight 164.99
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dichlorofluorobenzene
EINECSNumber (3)-4079
TSCA No