missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzylideneacetone, 97%

CAS: 61888-78-6 | C10H8Cl2O | 215.073 g/mol

$42.01 - $42.01

Chemical Identifiers

CAS 61888-78-6
Molecular Formula C10H8Cl2O
Molecular Weight (g/mol) 215.073
MDL Number MFCD00052852
InChI Key SAEQHTBHPNKKRX-NSCUHMNNSA-N
Synonym 2,4-dichlorobenzylideneacetone, 4-2,4-dichlorophenyl-3-buten-2-one, e-4-2,4-dichlorophenyl but-3-en-2-one, 3e-4-2,4-dichlorophenyl but-3-en-2-one, e-4-2,4-dichloro-phenyl-but-3-en-2-one, 4-2,4-dichlorophenyl-3-butene-2-one
PubChem CID 5849647
IUPAC Name (E)-4-(2,4-dichlorophenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=C(C=C(C=C1)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2527003
View Documents
Thermo Scientific Chemicals
B2527003
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

61888-78-6
215.073
SAEQHTBHPNKKRX-NSCUHMNNSA-N
5849647
CC(=O)C=CC1=C(C=C(C=C1)Cl)Cl
C10H8Cl2O
MFCD00052852
2,4-dichlorobenzylideneacetone, 4-2,4-dichlorophenyl-3-buten-2-one, e-4-2,4-dichlorophenyl but-3-en-2-one, 3e-4-2,4-dichlorophenyl but-3-en-2-one, e-4-2,4-dichloro-phenyl-but-3-en-2-one, 4-2,4-dichlorophenyl-3-butene-2-one
(E)-4-(2,4-dichlorophenyl)but-3-en-2-one

Specifications

61888-78-6
C10H8Cl2O
1 g
SAEQHTBHPNKKRX-NSCUHMNNSA-N
(E)-4-(2,4-dichlorophenyl)but-3-en-2-one
5849647
97%
83°C to 85°C
MFCD00052852
2,4-dichlorobenzylideneacetone, 4-2,4-dichlorophenyl-3-buten-2-one, e-4-2,4-dichlorophenyl but-3-en-2-one, 3e-4-2,4-dichlorophenyl but-3-en-2-one, e-4-2,4-dichloro-phenyl-but-3-en-2-one, 4-2,4-dichlorophenyl-3-butene-2-one
CC(=O)C=CC1=C(C=C(C=C1)Cl)Cl
215.073
215.08
2,4-Dichlorobenzylideneacetone
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only