missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2,4-Dichlorobenzyl)triphenylphosphonium Chloride 98.0+%, TCI America™
$83.89 - $208.78
Chemical Identifiers
| CAS | 2492-23-1 |
|---|---|
| Molecular Formula | C25H20Cl3P |
| Molecular Weight (g/mol) | 457.759 |
| MDL Number | MFCD00031641 |
| InChI Key | FWBSWSPGFNAXPP-UHFFFAOYSA-M |
| Synonym | 2,4-dichlorobenzyl triphenylphosphonium chloride, 2,4-dichlorobenzyltriphenylphosphonium chloride, phosphonium, 2,4-dichlorobenzyl triphenyl-, chloride, 2,4-dichlorophenyl methyl triphenylphosphonium chloride, phosphonium, 2,4-dichlorophenyl methyl triphenyl-, chloride, triphenyl 2,4-dichlorobenzyl phosphonium chloride, 2,4-dichlorophenyl methyl triphenylphosphanium chloride, phosphonium,4-dichlorobenzyl triphenyl-, chloride, phosphonium,4-dichlorophenyl methyl triphenyl-, chloride |
| PubChem CID | 196999 |
| IUPAC Name | (2,4-dichlorophenyl)methyl-triphenylphosphanium;chloride |
| SMILES | C1=CC=C(C=C1)[P+](CC2=C(C=C(C=C2)Cl)Cl)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-] |
Chemical Identifiers
| 2492-23-1 | |
| 457.759 | |
| FWBSWSPGFNAXPP-UHFFFAOYSA-M | |
| 196999 | |
| C1=CC=C(C=C1)[P+](CC2=C(C=C(C=C2)Cl)Cl)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-] |
| C25H20Cl3P | |
| MFCD00031641 | |
| 2,4-dichlorobenzyl triphenylphosphonium chloride, 2,4-dichlorobenzyltriphenylphosphonium chloride, phosphonium, 2,4-dichlorobenzyl triphenyl-, chloride, 2,4-dichlorophenyl methyl triphenylphosphonium chloride, phosphonium, 2,4-dichlorophenyl methyl triphenyl-, chloride, triphenyl 2,4-dichlorobenzyl phosphonium chloride, 2,4-dichlorophenyl methyl triphenylphosphanium chloride, phosphonium,4-dichlorobenzyl triphenyl-, chloride, phosphonium,4-dichlorophenyl methyl triphenyl-, chloride | |
| (2,4-dichlorophenyl)methyl-triphenylphosphanium;chloride |
Specifications
| 2492-23-1 | |
| White | |
| MFCD00031641 | |
| 2,4-dichlorobenzyl triphenylphosphonium chloride, 2,4-dichlorobenzyltriphenylphosphonium chloride, phosphonium, 2,4-dichlorobenzyl triphenyl-, chloride, 2,4-dichlorophenyl methyl triphenylphosphonium chloride, phosphonium, 2,4-dichlorophenyl methyl triphenyl-, chloride, triphenyl 2,4-dichlorobenzyl phosphonium chloride, 2,4-dichlorophenyl methyl triphenylphosphanium chloride, phosphonium,4-dichlorobenzyl triphenyl-, chloride, phosphonium,4-dichlorophenyl methyl triphenyl-, chloride | |
| C1=CC=C(C=C1)[P+](CC2=C(C=C(C=C2)Cl)Cl)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-] | |
| 457.759 | |
| 457.76 | |
| Crystalline Powder |
| 249°C | |
| C25H20Cl3P | |
| 5 g | |
| FWBSWSPGFNAXPP-UHFFFAOYSA-M | |
| (2,4-dichlorophenyl)methyl-triphenylphosphanium;chloride | |
| 196999 | |
| ≥98.0% (HPLC,T) | |
| (2,4-Dichlorobenzyl)triphenylphosphonium Chloride |
Safety and Handling
RTECSNumber : TA2200000
TSCA : No