missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichlorobenzyl Chloride 98.0+%, TCI America™
$39.14 - $238.60
Chemical Identifiers
| CAS | 94-99-5 |
|---|---|
| Molecular Formula | C7H5Cl3 |
| Molecular Weight (g/mol) | 195.47 |
| MDL Number | MFCD00000895 |
| InChI Key | IRSVDHPYXFLLDS-UHFFFAOYSA-N |
| Synonym | 2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro |
| PubChem CID | 7212 |
| IUPAC Name | 2,4-dichloro-1-(chloromethyl)benzene |
| SMILES | ClCC1=CC=C(Cl)C=C1Cl |
Chemical Identifiers
| 94-99-5 | |
| 195.47 | |
| IRSVDHPYXFLLDS-UHFFFAOYSA-N | |
| 7212 | |
| ClCC1=CC=C(Cl)C=C1Cl |
| C7H5Cl3 | |
| MFCD00000895 | |
| 2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro | |
| 2,4-dichloro-1-(chloromethyl)benzene |
Specifications
| 94-99-5 | |
| 248°C | |
| MFCD00000895 | |
| 3265 | |
| IRSVDHPYXFLLDS-UHFFFAOYSA-N | |
| 2,4-dichloro-1-(chloromethyl)benzene | |
| 7212 | |
| ≥98.0% (GC) | |
| 2,4-Dichlorobenzyl Chloride |
| Colorless | |
| C7H5Cl3 | |
| 25 g | |
| 2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro | |
| ClCC1=CC=C(Cl)C=C1Cl | |
| 195.47 | |
| 195.47 | |
| Liquid |
Safety and Handling
TSCA : Yes