missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl chloride, 98%

CAS: 94-99-5 | C7H5Cl3 | 195.47 g/mol

Supplier:  Thermo Scientific Chemicals A114350B

 View more versions of this product

Catalog No. AAA114350B


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dichlorobenzyl chloride
-3°C
248°C
Pungent
1.576
1000 g
387220
IRSVDHPYXFLLDS-UHFFFAOYSA-N
2,4-dichloro-1-(chloromethyl)benzene
7212
98%
94-99-5
1.408
190°C (374°F)
C7H5Cl3
MFCD00000895
UN3265
2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro
ClCC1=CC=C(Cl)C=C1Cl
195.47
195.48

Chemical Identifiers

94-99-5
195.47
IRSVDHPYXFLLDS-UHFFFAOYSA-N
7212
ClCC1=CC=C(Cl)C=C1Cl
C7H5Cl3
MFCD00000895
2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro
2,4-dichloro-1-(chloromethyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H332-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 202-381-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only