missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichlorobenzoyl Chloride 97.0+%, TCI America™
$44.28 - $232.95
Chemical Identifiers
| CAS | 89-75-8 |
|---|---|
| Molecular Formula | C7H3Cl3O |
| Molecular Weight (g/mol) | 209.45 |
| MDL Number | MFCD00000661 |
| InChI Key | CEOCVKWBUWKBKA-UHFFFAOYSA-N |
| Synonym | benzoyl chloride, 2,4-dichloro, 2,4-dichlorobenzoylchloride, benzoylchloride, 2,4-dichloro, ccris 8616, acmc-1cuf7, dsstox_cid_31176, dsstox_gsid_52603, 2,4-dichloro benzoylchloride, 2.4-dichlorobenzoyl chloride, ksc450c2p |
| PubChem CID | 66645 |
| IUPAC Name | 2,4-dichlorobenzoyl chloride |
| SMILES | ClC(=O)C1=CC=C(Cl)C=C1Cl |
Chemical Identifiers
| 89-75-8 | |
| 209.45 | |
| CEOCVKWBUWKBKA-UHFFFAOYSA-N | |
| 66645 | |
| ClC(=O)C1=CC=C(Cl)C=C1Cl |
| C7H3Cl3O | |
| MFCD00000661 | |
| benzoyl chloride, 2,4-dichloro, 2,4-dichlorobenzoylchloride, benzoylchloride, 2,4-dichloro, ccris 8616, acmc-1cuf7, dsstox_cid_31176, dsstox_gsid_52603, 2,4-dichloro benzoylchloride, 2.4-dichlorobenzoyl chloride, ksc450c2p | |
| 2,4-dichlorobenzoyl chloride |
Specifications
| 89-75-8 | |
| C7H3Cl3O | |
| 25 g | |
| benzoyl chloride, 2,4-dichloro, 2,4-dichlorobenzoylchloride, benzoylchloride, 2,4-dichloro, ccris 8616, acmc-1cuf7, dsstox_cid_31176, dsstox_gsid_52603, 2,4-dichloro benzoylchloride, 2.4-dichlorobenzoyl chloride, ksc450c2p | |
| ClC(=O)C1=CC=C(Cl)C=C1Cl | |
| 209.45 | |
| 209.45 | |
| Liquid |
| Yellow | |
| MFCD00000661 | |
| 3265 | |
| CEOCVKWBUWKBKA-UHFFFAOYSA-N | |
| 2,4-dichlorobenzoyl chloride | |
| 66645 | |
| ≥97.0% (T) | |
| 2,4-Dichlorobenzoyl Chloride |
Safety and Handling
EINECSNumber : (3)-0062&(3)-1409
RTECSNumber : DM6636733
TSCA : Yes