missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoyl Chloride 97.0+%, TCI America™

Catalog No. D034525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D034525G 25 g
D0345500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D034525G Supplier TCI America Supplier No. D034525G
Only null left

Chemical Identifiers

CAS 89-75-8
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00000661
InChI Key CEOCVKWBUWKBKA-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,4-dichloro, 2,4-dichlorobenzoylchloride, benzoylchloride, 2,4-dichloro, ccris 8616, acmc-1cuf7, dsstox_cid_31176, dsstox_gsid_52603, 2,4-dichloro benzoylchloride, 2.4-dichlorobenzoyl chloride, ksc450c2p
PubChem CID 66645
IUPAC Name 2,4-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC=C(Cl)C=C1Cl

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 209.45
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2,4-Dichlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
RTECSNumber DM6636733
TSCA Yes