missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichlorobenzotrifluoride 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 118°C |
| Quantity | 25 g |
| UN Number | 1760 |
| Formula Weight | 215.00 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,4-Dichlorobenzotrifluoride |
Safety and Handling
| EINECSNumber | (3)-3749 |
| RTECSNumber | CZ5566877 |
| TSCA | Yes |