missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoic Acid 97.0+%, TCI America™

Supplier:  TCI America D0337500G

 View more versions of this product

Catalog No. D0337500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,4-Dichlorobenzoic Acid
162°C
C7H4Cl2O2
500 g
ATCRIUVQKHMXSH-UHFFFAOYSA-N
2,4-dichlorobenzoic acid
5787
191.01
Crystalline Powder
50-84-0
White-Yellow
MFCD00002414
benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
C1=CC(=C(C=C1Cl)Cl)C(=O)O
191.007
CHEBI:30748
≥97.0% (GC,T)

Chemical Identifiers

50-84-0
191.007
ATCRIUVQKHMXSH-UHFFFAOYSA-N
5787
2,4-dichlorobenzoic acid
C7H4Cl2O2
MFCD00002414
benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
CHEBI:30748
C1=CC(=C(C=C1Cl)Cl)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1410&(3)-1424

RTECSNumber : DG6650000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions