missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoic acid, 98%

CAS: 50-84-0 | C7H4Cl2O2 | 191.01 g/mol

$43.95 - $43.95

Chemical Identifiers

CAS 50-84-0
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002414
InChI Key ATCRIUVQKHMXSH-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
PubChem CID 5787
ChEBI CHEBI:30748
IUPAC Name 2,4-dichlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC160900050
SDS View Documents
Thermo Scientific Chemicals
160900050
5 g Glass Bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-84-0
191.01
ATCRIUVQKHMXSH-UHFFFAOYSA-N
5787
2,4-dichlorobenzoic acid
C7H4Cl2O2
MFCD00002414
benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
CHEBI:30748
C1=CC(=C(C=C1Cl)Cl)C(=O)O

Specifications

50-84-0
100.0
White
Authentic
Glass Bottle
Cl2C6H3CO2H
5 g
benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
ATCRIUVQKHMXSH-UHFFFAOYSA-N
2,4-dichlorobenzoic acid
5787
191.01
Powder
97.5
160.0°C to 164.0°C
>200.0°C
97.5% min. (HPLC)
C7H4Cl2O2
MFCD00002414
09, 342
Solubility in water: 0.36g/L (15°C). Other solubilities: soluble in alcohol,ether and acetone,insoluble in heptane
C1=CC(=C(C=C1Cl)Cl)C(=O)O
191.01
CHEBI:30748
98%
2, 4-Dichlorobenzoic acid, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

EINECSNumber : 200-067-8

RTECSNumber : DG6650000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only