missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoic acid, 98%

Catalog No. AC160900050
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass Bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160900050 5 g Glass Bottle
AC160901000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160900050 Supplier Thermo Scientific Chemicals Supplier No. 160900050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50-84-0 | C7H4Cl2O2 | 191.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50-84-0
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002414
InChI Key ATCRIUVQKHMXSH-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
PubChem CID 5787
ChEBI CHEBI:30748
IUPAC Name 2,4-dichlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160.0°C to 164.0°C
Color White
Boiling Point >200.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula Cl2C6H3CO2H
Quantity 5 g
Beilstein 09, 342
Solubility Information Solubility in water: 0.36g/L (15°C). Other solubilities: soluble in alcohol,ether and acetone,insoluble in heptane
Formula Weight 191.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 4-Dichlorobenzoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 200-067-8
RTECSNumber DG6650000
TSCA TSCA

RUO – Research Use Only