missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoic acid, 98%

Catalog No. AAA123720B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA123720B 1000 g
AAA1237230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA123720B Supplier Thermo Scientific Chemicals Supplier No. A123720B
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-84-0 | C7H4Cl2O2 | 191.007 g/mol

2,4-Dichlorobenzoic acid is used as reagent during the synthesis of pyrimido[2?,1?:2,3]thiazolo[4,5-b]quinoxaline derivatives. It is also used as starting reagent during the synthesis of 1-(substituted)-1,4-dihydro-6-nitro-4-oxo-7-(sub-secondary amino)-quinoline-3-carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorobenzoic acid is used as reagent during the synthesis of pyrimido[2′,1′:2,3]thiazolo[4,5-b]quinoxaline derivatives. It is also used as starting reagent during the synthesis of 1-(substituted)-1,4-dihydro-6-nitro-4-oxo-7-(sub-secondary amino)-quinoline-3-carboxylic acids.

Solubility
Soluble in ethanol (100 mg/ml), water (0.48 mg/ml), alcohol, ether, and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 50-84-0
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
MDL Number MFCD00002414
InChI Key ATCRIUVQKHMXSH-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
PubChem CID 5787
ChEBI CHEBI:30748
IUPAC Name 2,4-dichlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)O

Specifications

Melting Point 159°C to 164°C
Quantity 1000 g
Beilstein 1868192
Solubility Information Soluble in ethanol (100mg/ml),water (0.48mg/ml),alcohol,ether,and acetone.
Formula Weight 191.01
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-067-8
RTECSNumber DG6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only