missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichlorobenzohydrazide 98.0+%, TCI America™
$57.75 - $196.36
Chemical Identifiers
| CAS | 5814-06-2 |
|---|---|
| Molecular Formula | C7H6Cl2N2O |
| Molecular Weight (g/mol) | 205.04 |
| MDL Number | MFCD00025113 |
| InChI Key | QOJQHOGSXXSMKX-UHFFFAOYSA-N |
| Synonym | 2,4-dichlorobenzhydrazide, 2,4-dichlorobenzoic acid hydrazide, 2,4-dichlorobenzenecarbohydrazide, acmc-1akyd, 2,4-dichlorobenzoylhydrazine, 2,4-dichlor-benzoyl-hydrazin, 2,4-dichlorobenzoic hydrazide, benzoic acid,2,4-dichloro-, hydrazide, benzoic acid, 2,4-dichloro-, hydrazide |
| PubChem CID | 138596 |
| IUPAC Name | 2,4-dichlorobenzohydrazide |
| SMILES | C1=CC(=C(C=C1Cl)Cl)C(=O)NN |
Chemical Identifiers
| 5814-06-2 | |
| 205.04 | |
| QOJQHOGSXXSMKX-UHFFFAOYSA-N | |
| 138596 | |
| C1=CC(=C(C=C1Cl)Cl)C(=O)NN |
| C7H6Cl2N2O | |
| MFCD00025113 | |
| 2,4-dichlorobenzhydrazide, 2,4-dichlorobenzoic acid hydrazide, 2,4-dichlorobenzenecarbohydrazide, acmc-1akyd, 2,4-dichlorobenzoylhydrazine, 2,4-dichlor-benzoyl-hydrazin, 2,4-dichlorobenzoic hydrazide, benzoic acid,2,4-dichloro-, hydrazide, benzoic acid, 2,4-dichloro-, hydrazide | |
| 2,4-dichlorobenzohydrazide |
Specifications
| 5814-06-2 | |
| White | |
| MFCD00025113 | |
| 2,4-dichlorobenzhydrazide, 2,4-dichlorobenzoic acid hydrazide, 2,4-dichlorobenzenecarbohydrazide, acmc-1akyd, 2,4-dichlorobenzoylhydrazine, 2,4-dichlor-benzoyl-hydrazin, 2,4-dichlorobenzoic hydrazide, benzoic acid,2,4-dichloro-, hydrazide, benzoic acid, 2,4-dichloro-, hydrazide | |
| C1=CC(=C(C=C1Cl)Cl)C(=O)NN | |
| 205.04 | |
| 205.04 | |
| Crystalline Powder |
| 169°C | |
| C7H6Cl2N2O | |
| 1 g | |
| QOJQHOGSXXSMKX-UHFFFAOYSA-N | |
| 2,4-dichlorobenzohydrazide | |
| 138596 | |
| ≥98.0% (GC,T) | |
| 2,4-Dichlorobenzohydrazide |
Safety and Handling
TSCA : No