Learn More
2,4-Dichlorobenzaldehyde, 98%
CAS: 874-42-0 | C7H4Cl2O | 175.008 g/mol
Supplier: Thermo Scientific Chemicals A1252536
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,4-Dichlorobenzaldehyde | |
| 69°C to 73°C | |
| 127°C (260°F) | |
| MFCD00003305 | |
| UN1759 | |
| Air Sensitive | |
| YSFBEAASFUWWHU-UHFFFAOYSA-N | |
| 2,4-dichlorobenzaldehyde | |
| 13404 | |
| 98% |
| 874-42-0 | |
| 233°C | |
| C7H4Cl2O | |
| 500 g | |
| 471601 | |
| benzaldehyde, 2,4-dichloro, unii-z08ql2124r, ccris 6013, 2,4-dichloro benzaldehyde, 2,4-dichloro-benzaldehyde, acmc-209qnb, dsstox_cid_2130, 2,4 dichloro benzaldehyde, 2,4-di-chlorobenzaldehyde, dsstox_rid_76500 | |
| C1=CC(=C(C=C1Cl)Cl)C=O | |
| 175.008 | |
| 175.01 |
Chemical Identifiers
| 874-42-0 | |
| 175.008 | |
| YSFBEAASFUWWHU-UHFFFAOYSA-N | |
| 13404 | |
| C1=CC(=C(C=C1Cl)Cl)C=O |
| C7H4Cl2O | |
| MFCD00003305 | |
| benzaldehyde, 2,4-dichloro, unii-z08ql2124r, ccris 6013, 2,4-dichloro benzaldehyde, 2,4-dichloro-benzaldehyde, acmc-209qnb, dsstox_cid_2130, 2,4 dichloro benzaldehyde, 2,4-di-chlorobenzaldehyde, dsstox_rid_76500 | |
| 2,4-dichlorobenzaldehyde |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 212-861-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only