missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzaldehyde, 98%

Catalog No. p-7020534
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1252522 100 g
AAA1252536 500 g
AAA125250E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1252522 Supplier Thermo Scientific Chemicals Supplier No. A1252522
Only null left

CAS: 874-42-0 | C7H4Cl2O | 175.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874-42-0
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003305
InChI Key YSFBEAASFUWWHU-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dichloro, unii-z08ql2124r, ccris 6013, 2,4-dichloro benzaldehyde, 2,4-dichloro-benzaldehyde, acmc-209qnb, dsstox_cid_2130, 2,4 dichloro benzaldehyde, 2,4-di-chlorobenzaldehyde, dsstox_rid_76500
PubChem CID 13404
IUPAC Name 2,4-dichlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)Cl)C=O

Specifications

Melting Point 69°C to 73°C
Boiling Point 233°C
Flash Point 127°C (260°F)
Quantity 100 g
UN Number UN1759
Beilstein 471601
Sensitivity Air Sensitive
Formula Weight 175.01
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzaldehyde
Hazard Category H314
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 212-861-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only