missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloroaniline 99.0+%, TCI America™

Supplier:  TCI America D0321500G

 View more versions of this product

Catalog No. D0321500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,4-Dichloroaniline
62°C
242°C
MFCD00007661
3442
KQCMTOWTPBNWDB-UHFFFAOYSA-N
2,4-dichloroaniline
11123
162.01
Crystalline Powder
554-00-7
White
C6H5Cl2N
500 g
2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german
C1=CC(=C(C=C1Cl)Cl)N
162.013
CHEBI:46635
≥99.0% (GC)

Chemical Identifiers

554-00-7
162.013
KQCMTOWTPBNWDB-UHFFFAOYSA-N
11123
2,4-dichloroaniline
C6H5Cl2N
MFCD00007661
2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german
CHEBI:46635
C1=CC(=C(C=C1Cl)Cl)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0261

RTECSNumber : BX2600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions