missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloroaniline 99.0+%, TCI America™

Catalog No. D032125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032125G 25 g
D0321100G 100 g
D0321500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D032125G Supplier TCI America Supplier No. D032125G

Chemical Identifiers

CAS 554-00-7
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007661
InChI Key KQCMTOWTPBNWDB-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german
PubChem CID 11123
ChEBI CHEBI:46635
IUPAC Name 2,4-dichloroaniline
SMILES C1=CC(=C(C=C1Cl)Cl)N

Specifications

Melting Point 62°C
Color White
Boiling Point 242°C
Quantity 25 g
UN Number 3442
Formula Weight 162.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloroaniline
EINECSNumber (3)-0261
RTECSNumber BX2600000
TSCA Yes