missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dichloroacetophenone 98.0+%, TCI America™

Catalog No. p-7126746
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D149025G 25 g
D1490500G 500 g
2 options

Catalog No. D149025G

Supplier: TCI America D149025G

Only null left

Chemical Identifiers

CAS 2234-16-4
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00000581
InChI Key XMCRWEBERCXJCH-UHFFFAOYSA-N
Synonym 2',4'-dichloroacetophenone, 1-2,4-dichlorophenyl ethanone, 2,4-dichloroacetophenone, ethanone, 1-2,4-dichlorophenyl, 1-2,4-dichlorophenyl ethan-1-one, p-chloro-2-chloroacetophenone, acetophenone, 2',4'-dichloro, unii-t7f2608h5h, 2',4'-dichloro phenyl ethanone, 1-2,4-dichloro-phenyl-ethanone
PubChem CID 16693
IUPAC Name 1-(2,4-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Cl)Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 189.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2′,4′-Dichloroacetophenone
EINECSNumber (3)-4370
TSCA Yes