accessibility menu, dialog, popup

UserName

H3COClCl

CAS RN 2234-16-4

IUPAC Name: 1-(2,4-dichlorophenyl)ethan-1-one
Synonyms: 1-(2,4-dichlorophenyl)ethanone
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: XMCRWEBERCXJCH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)