missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dichloroacetophenone, 97%

Catalog No. AC302432500
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302432500 250 g Glass bottle
AC302430250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC302432500 Supplier Thermo Scientific Chemicals Supplier No. 302432500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 2234-16-4 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2234-16-4
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000581
InChI Key XMCRWEBERCXJCH-UHFFFAOYSA-N
Synonym 2',4'-dichloroacetophenone, 1-2,4-dichlorophenyl ethanone, 2,4-dichloroacetophenone, ethanone, 1-2,4-dichlorophenyl, 1-2,4-dichlorophenyl ethan-1-one, p-chloro-2-chloroacetophenone, acetophenone, 2',4'-dichloro, unii-t7f2608h5h, 2',4'-dichloro phenyl ethanone, 1-2,4-dichloro-phenyl-ethanone
PubChem CID 16693
IUPAC Name 1-(2,4-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 29.0°C to 34.0°C
Color Brown-Yellow to Colorless
Density 1.3200g/mL
Boiling Point 242.0°C
Flash Point 131°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5635
Linear Formula Cl2C6H3COCH3
Quantity 250 g
Beilstein 07, II, 219
Specific Gravity 1.32
Solubility Information Solubility in water: ca. 200mg/l in water
Formula Weight 189.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2', 4'-Dichloroacetophenone, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-780-8
TSCA TSCA

RUO – Research Use Only