missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-N-methylbenzylamine 98.0+%, TCI America™

Catalog No. D51471G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51471G 1 g
D51475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51471G Supplier TCI America Supplier No. D51471G
Only null left

Chemical Identifiers

CAS 5013-77-4
Molecular Formula C8H9Cl2N
Molecular Weight (g/mol) 190.067
MDL Number MFCD00045185
InChI Key GUJXWKXDISDARD-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl methylamine, n-methyl-2,4-dichlorobenzylamine, benzenemethanamine, 2,4-dichloro-n-methyl, 2,4-dichlorophenyl methyl methyl amine, 2,4-dichloro-n-methylbenzylamine, 1-2,4-dichlorophenyl-n-methylmethanamine, 2,4-dichlorophenyl-n-methylmethanamine, n-2,4-dichlorobenzyl-n-methylamine, 1-2,4-dichlorophenyl-n-methyl-methanamine, 2,4-dichlorobenzyl methyl amine
PubChem CID 485424
IUPAC Name 1-(2,4-dichlorophenyl)-N-methylmethanamine
SMILES CNCC1=C(C=C(C=C1)Cl)Cl

Specifications

Color Yellow
Boiling Point 123°C
Quantity 1 g
UN Number 2735
Formula Weight 190.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4-Dichloro-N-methylbenzylamine