missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-nitrophenol, 98+%

Catalog No. p-7055841
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6452414 25 g
AAH6452422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6452414 Supplier Thermo Scientific Chemicals Supplier No. H6452414

CAS: 609-89-2 | C6H3Cl2NO3 | 207.994 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-89-2
Molecular Formula C6H3Cl2NO3
Molecular Weight (g/mol) 207.994
MDL Number MFCD00007101
InChI Key LYPMXMBQPXPNIQ-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-nitrophenol, phenol, 2,4-dichloro-6-nitro, 2,4-dichlor-6-nitrofenol, 4,6-dichloro-2-nitrophenol, 2,4-dichlor-6-nitrofenol czech, pubchem22918, acmc-1aunw, wln: wnr bq cg eg, 2-nitro-4,6-dichlorophenol
PubChem CID 11871
SMILES C1=C(C=C(C(=C1Cl)O)[N+](=O)[O-])Cl

Specifications

Melting Point 118°C to 120°C
Color Yellow
Quantity 25 g
UN Number UN2811
Beilstein 2050081
Formula Weight 207.99
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 2,4-Dichloro-6-nitrophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 210-202-2
RTECSNumber SL0350000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only