missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-morpholino-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7128480
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48141G 1 g
D48145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48141G Supplier TCI America Supplier No. D48141G
Only null left

Chemical Identifiers

CAS 6601-22-5
Molecular Formula C7H8Cl2N4O
Molecular Weight (g/mol) 235.068
MDL Number MFCD00460178
InChI Key UQAMDAUJTXFNAD-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-morpholino-1,3,5-triazine, 4-4,6-dichloro-1,3,5-triazin-2-yl morpholine, unii-ndh2046uk7, 2,4-dichloro-6-4-morpholinyl-1,3,5-triazine, 2,4-dichloro-6-morpholin-4-yl-1,3,5-triazine, 1,3,5-triazine, 2,4-dichloro-6-4-morpholinyl, 2,4-dichloro-6-morpholin-4-yl-1,3,5 triazine, pubchem21087, 2,4-dichlor-6-morpholin-4-yl-1,3,5-triazin, 2,6-dichloro-4-morpholino-1,3,5-triazine
PubChem CID 81076
IUPAC Name 4-(4,6-dichloro-1,3,5-triazin-2-yl)morpholine
SMILES C1COCCN1C2=NC(=NC(=N2)Cl)Cl

Specifications

Melting Point 161°C
Color White
Quantity 1 g
Formula Weight 235.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-6-morpholino-1,3,5-triazine